Product Details

SNP ID
rs7529714
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:153541424 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAGAGAATCCAAGCAGCAGGCA[C/T]AGCCTCCCTCTCACTGCCTTCTGCC
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002962.1 652 UTR 5 NP_002953.2
XM_011509860.2 652 Intron XP_011508162.1
XM_017002029.1 652 Missense Mutation ATG,GTG M24V XP_016857518.1
XM_017002030.1 652 Missense Mutation ATG,GTG M1V XP_016857519.1
XM_017002031.1 652 Intron XP_016857520.1
XM_017002032.1 652 Intron XP_016857521.1
Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details