Product Details

SNP ID
rs17272847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49476141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGGCCTGATCCTGTTTTCTCCC[A/G]CAGTCTGACTACCTGCTTCAAGATT
Phenotype
MIM: 613358 MIM: 600007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH16A1 PubMed Links

Gene Details

Gene
ALDH16A1
Gene Name
aldehyde dehydrogenase 16 family member A1
There are no transcripts associated with this gene.

Gene
FLT3LG
Gene Name
fms related tyrosine kinase 3 ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204502.1 274 Intron NP_001191431.1
NM_001204503.1 274 Intron NP_001191432.1
NM_001278637.1 274 UTR 5 NP_001265566.1
NM_001278638.1 274 Intron NP_001265567.1
NM_001459.3 274 Intron NP_001450.2
XM_005258680.4 274 Intron XP_005258737.3
XM_005258681.4 274 Intron XP_005258738.3
XM_005258682.4 274 Intron XP_005258739.3
XM_005258683.4 274 Intron XP_005258740.3
XM_006723116.3 274 Intron XP_006723179.2
XM_011526675.2 274 Intron XP_011524977.1
XM_011526676.2 274 Intron XP_011524978.1
XM_011526677.2 274 Intron XP_011524979.1
XM_011526678.2 274 Intron XP_011524980.1
XM_011526680.2 274 Intron XP_011524982.1
XM_011526682.2 274 Intron XP_011524984.1
XM_017026532.1 274 Intron XP_016882021.1
XM_017026533.1 274 Intron XP_016882022.1
XM_017026534.1 274 Intron XP_016882023.1
XM_017026535.1 274 UTR 5 XP_016882024.1

View Full Product Details