Product Details

SNP ID
rs35804663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:105174323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGCGCAAGTGAGTTATGCTTC[A/G]GATTCTGCGCCTCTATGTTCGTTTG
Phenotype
MIM: 606231 MIM: 615748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APPL2 PubMed Links

Gene Details

Gene
APPL2
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251904.1 3319 Silent Mutation NP_001238833.1
NM_001251905.1 3319 Silent Mutation NP_001238834.1
NM_018171.3 3319 Silent Mutation NP_060641.2
XM_006719472.1 3319 Silent Mutation XP_006719535.1
XM_011538530.2 3319 Silent Mutation XP_011536832.1
XM_011538531.2 3319 Silent Mutation XP_011536833.1
XM_011538532.2 3319 Silent Mutation XP_011536834.1
XM_017019551.1 3319 Silent Mutation XP_016875040.1
XM_017019552.1 3319 Silent Mutation XP_016875041.1
XM_017019553.1 3319 Silent Mutation XP_016875042.1
XM_017019554.1 3319 Silent Mutation XP_016875043.1
Gene
KIAA1033
Gene Name
KIAA1033
There are no transcripts associated with this gene.

View Full Product Details