Product Details

SNP ID
rs10864654
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:231020283 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGGATGCTTTGGATTTCCAGTTTTG[A/G]GAACACCTTAATTGTGTTATGCCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM89A PubMed Links

Gene Details

Gene
FAM89A
Gene Name
family with sequence similarity 89 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198552.2 Intron NP_940954.1
Gene
MIR1182
Gene Name
microRNA 1182
There are no transcripts associated with this gene.

View Full Product Details