Product Details

SNP ID
rs56266102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167000170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGTTAATACTTAATTTCTTATT[C/T]ATTAAAAACCGTCTTAAAAATAGTT
Phenotype
MIM: 605392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFR1OP PubMed Links

Gene Details

Gene
FGFR1OP
Gene Name
FGFR1 oncogene partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278690.1 Intron NP_001265619.1
NM_007045.3 Intron NP_008976.1
NM_194429.2 Intron NP_919410.1
Gene
MIR3939
Gene Name
microRNA 3939
There are no transcripts associated with this gene.

View Full Product Details