Product Details

SNP ID
rs4802566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49129052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCAGGGGTGGAGAATGGAAACC[C/T]ATAGTTGACTGGGGCTGTTCTGAGT
Phenotype
MIM: 603144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
There are no transcripts associated with this gene.

Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 Intron NP_003651.1
XM_017027407.1 Intron XP_016882896.1

View Full Product Details