Product Details

SNP ID
rs61739276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149003913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGTTCTTTTTGTCCCCACTGG[G/T]GGCGAAGTAGATGGTGGGGAAGCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDIA4 PubMed Links

Gene Details

Gene
PDIA4
Gene Name
protein disulfide isomerase family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004911.4 2101 Missense Mutation ACC,CCC T607P NP_004902.1
XM_006716185.2 2101 Missense Mutation ACC,CCC T608P XP_006716248.1

View Full Product Details