Product Details

SNP ID
rs61743784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29780104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCTTCCTCCCCTCTTCCTCAG[A/T]CTTCAGGTGCGCTATGGCAAGGTGT
Phenotype
MIM: 603213
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
ZG16
Gene Name
zymogen granule protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152338.3 342 Silent Mutation GGA,GGT G63G NP_689551.2
XM_011545921.2 342 Silent Mutation GGA,GGT G63G XP_011544223.1

View Full Product Details