Product Details

SNP ID
rs12280564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36592402 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTAAAATATTTTCAAAATGTATA[G/T]GGTCTAGAATGACTTTATTTATCAT
Phenotype
MIM: 179616
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf74 PubMed Links

Gene Details

Gene
C11orf74
Gene Name
chromosome 11 open reading frame 74
There are no transcripts associated with this gene.

Gene
RAG2
Gene Name
recombination activating gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000536.3 1972 UTR 3 NP_000527.2
NM_001243785.1 1972 UTR 3 NP_001230714.1
NM_001243786.1 1972 UTR 3 NP_001230715.1

View Full Product Details