Product Details

SNP ID
rs76501895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36856552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAGTGAGCTTTTAAAATCCCC[A/G]TTCCTACCCAGTCCAGCCAAATTAT
Phenotype
MIM: 616642 MIM: 601301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf89 PubMed Links

Gene Details

Gene
C6orf89
Gene Name
chromosome 6 open reading frame 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286635.1 Intron NP_001273564.1
NM_001286636.1 Intron NP_001273565.1
NM_001286637.1 Intron NP_001273566.1
NM_152734.3 Intron NP_689947.2
XM_011514377.2 Intron XP_011512679.1
XM_017010432.1 Intron XP_016865921.1
XM_017010433.1 Intron XP_016865922.1
XM_017010434.1 Intron XP_016865923.1
Gene
PPIL1
Gene Name
peptidylprolyl isomerase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016059.4 Intron NP_057143.1

View Full Product Details