Product Details

SNP ID
rs35036798
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.16:75478724 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GGCAGCACCAGATCAAGGGCGAGGT[C/T]GCGCTGCTCGTCCTCAGAGTACACA
Phenotype
MIM: 605294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST6 PubMed Links

Gene Details

Gene
CHST6
Gene Name
carbohydrate sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021615.4 1900 Missense Mutation AAC,GAC N369D NP_067628.1
XM_005255955.4 1900 Missense Mutation AAC,GAC N369D XP_005256012.1
XM_011523085.2 1900 Missense Mutation AAC,GAC N369D XP_011521387.1

View Full Product Details