Product Details
- SNP ID
-
rs79770432
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:56462901 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCTCACATCTTCCAAGAACTGCT[C/T]GGTTGATACTTATCTTATAGAAGTG
- Phenotype
-
MIM: 113810
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DST
PubMed Links
Gene Details
- Gene
- DST
- Gene Name
- dystonin
View Full Product Details