Product Details

SNP ID
rs36091974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39184058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGAGGTGGGAGCCTGGGTTCCT[C/G]GGTGGCTCTGCCTGCTCGGTGACTG
Phenotype
MIM: 609777
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XIRP1 PubMed Links

Gene Details

Gene
XIRP1
Gene Name
xin actin binding repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198621.2 5192 UTR 3 NP_001185550.1
NM_194293.2 5192 Silent Mutation CCC,CCG P1796P NP_919269.2
XM_005264909.3 5192 Silent Mutation CCC,CCG P1796P XP_005264966.1

View Full Product Details