Product Details
- SNP ID
-
rs34119369
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:145573 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTGCTAATTATCCTGTTTTCTGT[A/G]TCCAGACAAAGACTACAAGAATTTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB126
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77783981] are located under a probe and SNP(s) [rs77106211] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEFB126
- Gene Name
- defensin beta 126
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030931.3 |
293 |
Missense Mutation |
ATC,GTC |
I73V |
NP_112193.1 |
View Full Product Details