Product Details

SNP ID
rs34543561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:93829499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATGTTATCCCGTCGTTCAACAA[C/T]GACAAGAAGCTGTCCAAATATGAGA
Phenotype
MIM: 601461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATOH1 PubMed Links

Gene Details

Gene
ATOH1
Gene Name
atonal bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005172.1 573 Silent Mutation AAC,AAT N191N NP_005163.1

View Full Product Details