Product Details

SNP ID
hCV25642096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183818161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCACAGGGTCAGCCACCCCTT[C/T]ACCGACTCCCCAGGGGCTGCTGCTC
Phenotype
MIM: 610593 MIM: 607858 MIM: 613373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP6D1 PubMed Links

Gene Details

Gene
MAP6D1
Gene Name
MAP6 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024871.2 Intron NP_079147.1
Gene
PARL
Gene Name
presenilin associated rhomboid like
There are no transcripts associated with this gene.

Gene
YEATS2
Gene Name
YEATS domain containing 2
There are no transcripts associated with this gene.

View Full Product Details