Product Details

SNP ID
rs11844594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77377471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCGGCCCAGATGAAGATGGAGAGC[C/T]GGAGCCTGAGAGGCCTGAACTGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAMD15 PubMed Links

Gene Details

Gene
SAMD15
Gene Name
sterile alpha motif domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010860.1 53 Missense Mutation CCG,CTG P18L NP_001010860.1
Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 53 Intron NP_998766.1
XM_017021223.1 53 Intron XP_016876712.1
XM_017021224.1 53 Intron XP_016876713.1
XM_017021225.1 53 Intron XP_016876714.1

View Full Product Details