Product Details

SNP ID
rs28637549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45800370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCCCCATCTCTGCCCACCCTGG[C/T]GAGTTGCCCAGCCCAACCAGGCTTG
Phenotype
MIM: 609857 MIM: 607548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMWD PubMed Links

Gene Details

Gene
DMWD
Gene Name
dystrophia myotonica, WD repeat containing
There are no transcripts associated with this gene.

Gene
RSPH6A
Gene Name
radial spoke head 6 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030785.3 Intron NP_110412.1
XM_011527351.1 Intron XP_011525653.1

View Full Product Details