Product Details

SNP ID
rs28483271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86048564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTGACTTAGAAGACTGTGCAGA[A/T]CCAGGACTCTGAGAGTGGGGAGAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PSKH2 PubMed Links

Gene Details

Gene
PSKH2
Gene Name
protein serine kinase H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033126.1 1056 Silent Mutation GGA,GGT G352G NP_149117.1
XM_017013929.1 1056 Silent Mutation GGA,GGT G471G XP_016869418.1

View Full Product Details