Product Details

SNP ID
rs34896991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93101432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTCTAAGTTGACCTAAAAAAG[T/C]GGGAGTGATGGGTATTGTGATTTTT
Phenotype
MIM: 610456
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SAMD9 PubMed Links

Gene Details

Gene
SAMD9
Gene Name
sterile alpha motif domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193307.1 4835 Missense Mutation ACT,GCT T1556A NP_001180236.1
NM_017654.3 4835 Missense Mutation ACT,GCT T1556A NP_060124.2

View Full Product Details