Product Details

SNP ID
hCV25643514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143935401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAGACTGGACAGCCGACTCCAC[A/G]CTCCCATGTATTTCTTTCTCACCAA
Phenotype
MIM: 608497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012369.2 169 Intron NP_036501.2
Gene
OR2F2
Gene Name
olfactory receptor family 2 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004685.1 169 Missense Mutation ACT,GCT T57A NP_001004685.1

View Full Product Details