Product Details

SNP ID
rs16980994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15949307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCACCTTTGTGGCTTGTGCCAA[A/C]CAGATGTTCTTCTCCTTCATGTTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10H4 PubMed Links

Gene Details

Gene
OR10H4
Gene Name
olfactory receptor family 10 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004465.1 300 Missense Mutation AAA,AAC K100N NP_001004465.1

View Full Product Details