Product Details

SNP ID
rs185729577
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1348000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGGTCCTCCCGGAGCTTTCGG[G/T]TTATAGTGGAACCCAGTAAACACGT
Phenotype
MIM: 606801 MIM: 614632
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links
Additional Information
For this assay, SNP(s) [rs115755535] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317934.1 Intron NP_001304863.1
NM_001317935.1 Intron NP_001304864.1
NM_020894.3 Intron NP_065945.2
XM_017008490.1 Intron XP_016863979.1
XM_017008491.1 Intron XP_016863980.1
XM_017008492.1 Intron XP_016863981.1
XM_017008493.1 Intron XP_016863982.1
XM_017008494.1 Intron XP_016863983.1
XM_017008495.1 Intron XP_016863984.1
XM_017008496.1 Intron XP_016863985.1
XM_017008497.1 Intron XP_016863986.1
XM_017008498.1 Intron XP_016863987.1
XM_017008499.1 Intron XP_016863988.1
XM_017008500.1 Intron XP_016863989.1

View Full Product Details