Product Details

SNP ID
rs35430846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46498209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGATGCAATACCAGCAGTGGAC[C/T]TTCGCCAGGTGAGGGGGAATGTGAT
Phenotype
MIM: 607181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTP3 PubMed Links

Gene Details

Gene
RTP3
Gene Name
receptor transporter protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031440.1 215 Silent Mutation ACC,ACT T49T NP_113628.1

View Full Product Details