Product Details
- SNP ID
-
rs62001054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142715735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTACTATTCAATGAATGTTGAGG[C/T]GACTGATAAGGGAGATGTTCCTGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRBV27
PubMed Links
Gene Details
- Gene
- TRBV27
- Gene Name
- T cell receptor beta variable 27
There are no transcripts associated with this gene.
View Full Product Details