Product Details

SNP ID
rs116198115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37211131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGCAATTTAAAAACAATAAGAC[C/T]GGAAAATATTACTCCAAGATATGCT
Phenotype
MIM: 612828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEBPZ PubMed Links

Gene Details

Gene
CEBPZ
Gene Name
CCAAT/enhancer binding protein zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005760.2 Intron NP_005751.2
Gene
CEBPZOS
Gene Name
CEBPZ opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322373.1 Intron NP_001309302.1
NM_001322374.1 Intron NP_001309303.1
NM_001322375.1 Intron NP_001309304.1
NM_001322376.1 Intron NP_001309305.1
XM_017003106.1 Intron XP_016858595.1
XM_017003107.1 Intron XP_016858596.1
XM_017003108.1 Intron XP_016858597.1
XM_017003109.1 Intron XP_016858598.1

View Full Product Details