Product Details

SNP ID
hCV25645283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216039228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAAGTACAGAAGCATATCCT[C/T]AAGATGTGAAGGCACTGGGGGATGG
Phenotype
MIM: 605843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PECR PubMed Links

Gene Details

Gene
PECR
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018441.5 1034 UTR 3 NP_060911.2

View Full Product Details