Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282856.1 | 234 | Intron | NP_001269785.1 | ||
NM_001282858.1 | 234 | Intron | NP_001269787.1 | ||
NM_001282860.1 | 234 | Intron | NP_001269789.1 | ||
NM_001282861.1 | 234 | Intron | NP_001269790.1 | ||
NM_032292.5 | 234 | Intron | NP_115668.4 | ||
XM_005245284.3 | 234 | Intron | XP_005245341.1 | ||
XM_005245286.3 | 234 | Intron | XP_005245343.1 | ||
XM_006711393.3 | 234 | Intron | XP_006711456.1 | ||
XM_006711394.3 | 234 | Intron | XP_006711457.1 | ||
XM_011509658.2 | 234 | Intron | XP_011507960.1 | ||
XM_011509659.2 | 234 | Intron | XP_011507961.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152280.4 | 234 | UTR 5 | NP_689493.3 | ||
XM_005245014.2 | 234 | UTR 5 | XP_005245071.1 | ||
XM_017000759.1 | 234 | UTR 5 | XP_016856248.1 |