Product Details
- SNP ID
-
rs115072733
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:39383902 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCCTTTTCCGCGCCCCAGGGTGC[G/T]CTCAGGATTTAGGATGCGGCGGGAG
- Phenotype
-
MIM: 610819
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A38
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116056544] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC25A38
- Gene Name
- solute carrier family 25 member 38
View Full Product Details