Product Details

SNP ID
rs74952975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122909418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGGCCAGACTTCTGCCTCCT[A/G]TGGCATTGATCCTCTCTCCTGGGCC
Phenotype
MIM: 609298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEMA5B PubMed Links

Gene Details

Gene
SEMA5B
Gene Name
semaphorin 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031702.3 4618 UTR 3 NP_001026872.2
NM_001256346.1 4618 UTR 3 NP_001243275.1
NM_001256347.1 4618 UTR 3 NP_001243276.1
NM_001256348.1 4618 UTR 3 NP_001243277.1
XM_017006638.1 4618 UTR 3 XP_016862127.1
XM_017006639.1 4618 Intron XP_016862128.1

View Full Product Details