Product Details

SNP ID
rs35903236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53866148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGAACAAGCAGTTGGTCATCAG[C/T]GAATTGCTGTTTATGTTCCTGCCAG
Phenotype
MIM: 606704 MIM: 607705
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR75 PubMed Links

Gene Details

Gene
GPR75
Gene Name
G protein-coupled receptor 75
There are no transcripts associated with this gene.

Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
There are no transcripts associated with this gene.

Gene
PSME4
Gene Name
proteasome activator subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014614.2 5565 Missense Mutation ACT,GCT T1825A NP_055429.2
XM_006711969.1 5565 Missense Mutation ACT,GCT T983A XP_006712032.1
XM_011532705.2 5565 Missense Mutation ACT,GCT T1851A XP_011531007.1
XM_011532706.2 5565 Missense Mutation ACT,GCT T1847A XP_011531008.1
XM_011532707.2 5565 Missense Mutation ACT,GCT T1829A XP_011531009.1
XM_011532709.2 5565 Intron XP_011531011.1

View Full Product Details