Product Details

SNP ID
rs73266403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28908962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAAACCAACAAAATCTGGACCT[C/T]TGGCTCATGGCCTGGAGAAAATCCC
Phenotype
MIM: 616157
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033561.1 Intron NP_001028733.1
NM_001290131.1 Intron NP_001277060.1
NM_020889.2 Intron NP_065940.1
XM_005258015.3 Intron XP_005258072.1
XM_005258016.3 Intron XP_005258073.1
XM_011525078.2 Intron XP_011523380.1
XM_011525079.2 Intron XP_011523381.1
XM_011525082.2 Intron XP_011523384.1
XM_011525083.2 Intron XP_011523385.1
XM_017024903.1 Intron XP_016880392.1
XM_017024904.1 Intron XP_016880393.1

View Full Product Details