Product Details

SNP ID
rs149573883
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:153390545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTCGGTCAACATGATGCCCAC[A/G]GACTTGCCCCACCTGAAAAACAGAA
Phenotype
MIM: 123885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A8 PubMed Links

Gene Details

Gene
S100A8
Gene Name
S100 calcium binding protein A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319196.1 133 Silent Mutation TCC,TCT S21S NP_001306125.1
NM_001319197.1 133 Silent Mutation TCC,TCT S20S NP_001306126.1
NM_001319198.1 133 Silent Mutation TCC,TCT S5S NP_001306127.1
NM_001319201.1 133 UTR 5 NP_001306130.1
NM_002964.4 133 UTR 5 NP_002955.2

View Full Product Details