Product Details

SNP ID
rs116130181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:156763024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTAAGGATGGAGATAACGCATA[C/T]GTTCTCTAATAGAATCAGCCACTGC
Phenotype
MIM: 608452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDGFC PubMed Links

Gene Details

Gene
PDGFC
Gene Name
platelet derived growth factor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016205.2 1368 UTR 3 NP_057289.1
XM_011532124.2 1368 UTR 3 XP_011530426.1
XM_017008455.1 1368 UTR 3 XP_016863944.1
XM_017008456.1 1368 UTR 3 XP_016863945.1

View Full Product Details