Product Details

SNP ID
rs34647773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102446716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGTCCCCGCATGAGCGCATGCA[C/T]CCCTACATCGAGCTGGCCTGGGGCT
Phenotype
MIM: 613302 MIM: 610929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 574 Silent Mutation CAC,CAT H143H NP_001119812.1
NM_001271818.1 574 Silent Mutation CAC,CAT H143H NP_001258747.1
NM_001271819.1 574 Silent Mutation CAC,CAT H66H NP_001258748.1
NM_032831.3 574 Silent Mutation CAC,CAT H143H NP_116220.1

View Full Product Details