Product Details

SNP ID
rs142753905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48692994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAGAAAAATTATGTGTT[C/T]TTTTAAAGAAGTTTTTTTCTCCTCT
Phenotype
MIM: 143055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNT1 PubMed Links

Gene Details

Gene
CCNT1
Gene Name
cyclin T1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001240.3 2379 UTR 3 NP_001231.2
NM_001277842.1 2379 UTR 3 NP_001264771.1
XM_017020197.1 2379 UTR 3 XP_016875686.1

View Full Product Details