Product Details

SNP ID
rs143217896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103080230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGGCCATCCTGGGGAAGGCAA[C/G]AGGAAGTCAGTAGTGCAAAGTAGGG
Phenotype
MIM: 300316
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NXF3 PubMed Links

Gene Details

Gene
NXF3
Gene Name
nuclear RNA export factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022052.1 Intron NP_071335.1

View Full Product Details