Product Details

SNP ID
rs41306275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102513793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAGCAATGGAAGCACCCATCA[G/T]AGCAGGTTAAAACAGGCTTATCCAG
Phenotype
MIM: 300939
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMSB15A PubMed Links

Gene Details

Gene
TMSB15A
Gene Name
thymosin beta 15a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021992.2 555 UTR 3 NP_068832.1

View Full Product Details