Product Details

SNP ID
rs189998491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2867415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTCAAAGAGCTCAGGAACGG[A/G]CAGGACATGGTGAGTGCAGGGCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
There are no transcripts associated with this gene.

Gene
ZNF556
Gene Name
zinc finger protein 556
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300843.1 81 UTR 5 NP_001287772.1
NM_024967.2 81 UTR 5 NP_079243.1

View Full Product Details