Product Details

SNP ID
rs796576397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7311905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGCTCTGGCCCGGTCCTAAG[C/T]TGGACTCCTCCTACACAATTTATTT
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143760.1 Intron NP_001137232.1
NM_001143761.1 Intron NP_001137233.1
NM_001143762.1 Intron NP_001137234.1
NM_001970.4 Intron NP_001961.1
XM_005256509.2 Intron XP_005256566.1
XM_011523710.2 Intron XP_011522012.1
XM_011523711.2 Intron XP_011522013.1
XM_011523712.2 Intron XP_011522014.1
XM_011523713.2 Intron XP_011522015.1
XM_017024300.1 Intron XP_016879789.1
XM_017024301.1 Intron XP_016879790.1
Gene
GPS2
Gene Name
G protein pathway suppressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004489.4 Intron NP_004480.1
Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details