Product Details

SNP ID
rs62619837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7075358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACAGAAAGCCAGGGTGCACTC[A/G]CTGGAAGCCGGTCGCATAGTCTGTT
Phenotype
MIM: 605999
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC10A PubMed Links

Gene Details

Gene
CLEC10A
Gene Name
C-type lectin domain family 10 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256412.1 Intron XP_005256469.1
XM_011523613.1 Intron XP_011521915.1
XM_011523614.2 Intron XP_011521916.1
XM_011523615.1 Intron XP_011521917.1

View Full Product Details