Product Details

SNP ID
rs17005979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27276739 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGGCTCAGATCATGTCTACTG[G/T]GAAGGAAGCAGCACACCGGCTGTCC
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 367 Missense Mutation GGG,GTG G4V NP_001290056.1
NM_001303128.1 367 Missense Mutation GGG,GTG G4V NP_001290057.1
NM_173650.2 367 Missense Mutation GGG,GTG G4V NP_775921.1
XM_006711995.3 367 Missense Mutation GGG,GTG G4V XP_006712058.1
XM_017003872.1 367 Missense Mutation GGG,GTG G4V XP_016859361.1
XM_017003873.1 367 Missense Mutation GGG,GTG G4V XP_016859362.1
XM_017003874.1 367 Missense Mutation GGG,GTG G4V XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318949.1 367 Intron NP_001305878.1
NM_001318950.1 367 Intron NP_001305879.1
NM_001318951.1 367 Intron NP_001305880.1
NM_003459.4 367 Intron NP_003450.2
XM_005264547.1 367 Intron XP_005264604.1
XM_005264548.3 367 Intron XP_005264605.1
XM_006712100.2 367 Intron XP_006712163.1
XM_011533102.2 367 Intron XP_011531404.1
XM_011533103.2 367 Intron XP_011531405.1
XM_017004873.1 367 Intron XP_016860362.1
XM_017004874.1 367 Intron XP_016860363.1
XM_017004875.1 367 Intron XP_016860364.1
Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details