Product Details

SNP ID
rs79177795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:644748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGACCCCAGAGGGTGCAGCCCT[C/G]TGTAACCTCACCTGGAGGTGCTGAG
Phenotype
MIM: 602635 MIM: 126452
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEAF1 PubMed Links

Gene Details

Gene
DEAF1
Gene Name
DEAF1, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293634.1 Intron NP_001280563.1
NM_021008.3 Intron NP_066288.2
XM_011519842.2 Intron XP_011518144.1
Gene
DRD4
Gene Name
dopamine receptor D4
There are no transcripts associated with this gene.

View Full Product Details