Product Details

SNP ID
rs4233368
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:161227454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTATGGGGAGGAATGGGCCGTAAC[A/C]TGATTCTCTGTGTCTTTTCTGAATG
Phenotype
MIM: 107670 MIM: 147139 MIM: 603881
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOA2 PubMed Links

Gene Details

Gene
APOA2
Gene Name
apolipoprotein A2
There are no transcripts associated with this gene.

Gene
FCER1G
Gene Name
Fc fragment of IgE receptor Ig
There are no transcripts associated with this gene.

Gene
MIR5187
Gene Name
microRNA 5187
There are no transcripts associated with this gene.

Gene
NR1I3
Gene Name
nuclear receptor subfamily 1 group I member 3
There are no transcripts associated with this gene.

Gene
TOMM40L
Gene Name
translocase of outer mitochondrial membrane 40 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286373.1 Intron NP_001273302.1
NM_001286374.1 Intron NP_001273303.1
NM_032174.5 Intron NP_115550.2
XM_006711572.2 Intron XP_006711635.1
XM_011510057.2 Intron XP_011508359.1
XM_017002481.1 Intron XP_016857970.1
XM_017002482.1 Intron XP_016857971.1

View Full Product Details