Product Details

SNP ID
rs56169224
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:56618759 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATTTTTTTTTCATACAACCCTGAGC[A/G]TTTGCTACATTCCTTTTTCTATTAA
Phenotype
MIM: 156351 MIM: 156358 MIM: 156360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MT1E PubMed Links

Gene Details

Gene
MT1E
Gene Name
metallothionein 1E
There are no transcripts associated with this gene.

Gene
MT1L
Gene Name
metallothionein 1L (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
MT2A
Gene Name
metallothionein 2A
There are no transcripts associated with this gene.

View Full Product Details