Product Details

SNP ID
rs61737594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95027042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAGGCCACAGGCCTCAGCTCCC[G/T]CTTGCCAATGATTTGATGACAAATC
Phenotype
MIM: 616581
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM4E PubMed Links

Gene Details

Gene
KDM4E
Gene Name
lysine demethylase 4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161630.1 1785 Silent Mutation CCG,CCT P495P NP_001155102.1

View Full Product Details