Product Details

SNP ID
rs61741859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21155434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGAATCCTGCACTGTAGGAGC[C/T]CACAATCAGCGAGGCACAGACCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AU1 PubMed Links

Gene Details

Gene
OR5AU1
Gene Name
olfactory receptor family 5 subfamily AU member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004731.1 592 Missense Mutation AGC,GGC S198G NP_001004731.1

View Full Product Details