Product Details

SNP ID
rs61740726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68302704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGATGGAACATGTGGTGGAGAAG[A/G]CAACTGTCCAGGGGGATTTGTTGGG
Phenotype
MIM: 613813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDM1 PubMed Links

Gene Details

Gene
MDM1
Gene Name
Mdm1 nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205028.1 1916 Missense Mutation NP_001191957.1
NM_001205029.1 1916 Intron NP_001191958.1
NM_017440.4 1916 Missense Mutation NP_059136.2
NM_020128.2 1916 Intron NP_064513.1
XM_005269026.1 1916 Missense Mutation XP_005269083.1
XM_011538569.1 1916 Intron XP_011536871.1
XM_017019646.1 1916 Missense Mutation XP_016875135.1
XM_017019647.1 1916 Missense Mutation XP_016875136.1
XM_017019648.1 1916 Missense Mutation XP_016875137.1
XM_017019649.1 1916 Missense Mutation XP_016875138.1
XM_017019650.1 1916 Missense Mutation XP_016875139.1

View Full Product Details