Product Details

SNP ID
rs77525146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16789052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTGGGGGACTCTGAGGGGCACT[C/T]GGCTCCCTGTCCCTGCCCCGCTGCC
Phenotype
MIM: 612686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC283278 PubMed Links

Gene Details

Gene
LOC283278
Gene Name
pleckstrin homology domain-containing family A member 6
There are no transcripts associated with this gene.

Gene
PLEKHA7
Gene Name
pleckstrin homology domain containing A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175058.4 3416 UTR 3 NP_778228.3

View Full Product Details