Product Details

SNP ID
rs111398015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68295208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTAACACAGTAAGCAATAAAG[A/T]AAAATTATGCCAATGTTTCCTTAGA
Phenotype
MIM: 613813
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MDM1 PubMed Links

Gene Details

Gene
MDM1
Gene Name
Mdm1 nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205028.1 2219 UTR 3 NP_001191957.1
NM_001205029.1 2219 Intron NP_001191958.1
NM_017440.4 2219 UTR 3 NP_059136.2
NM_020128.2 2219 Intron NP_064513.1
XM_005269026.1 2219 UTR 3 XP_005269083.1
XM_011538569.1 2219 Intron XP_011536871.1
XM_017019646.1 2219 UTR 3 XP_016875135.1
XM_017019647.1 2219 UTR 3 XP_016875136.1
XM_017019648.1 2219 UTR 3 XP_016875137.1
XM_017019649.1 2219 UTR 3 XP_016875138.1
XM_017019650.1 2219 UTR 3 XP_016875139.1

View Full Product Details